missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABHD7 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£161.00 - £389.00
Specifications
| Antigen | ABHD7 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17920383
|
Invitrogen™
PA5145426 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17900133
![]() |
Invitrogen™
PA5145425 |
200 μL |
£389.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ABHD7 gene ontology (GO) annotation includes hydrolase activity and membrane.Specifications
| ABHD7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q6IE26, Q8IUS5 | |
| 253152, 384214 | |
| Recombinant Mouse EPHX4 protein, Lys36-Asp359 (Accession #Q6IE26). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| ABHD7; abhydrolase domain containing 7; abhydrolase domain-containing protein 7; EH4; EPHX4; Ephxrp; epoxide hydrolase 4; Epoxide hydrolase-related protein; Gm1382 | |
| EPHX4 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| EPHX4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title