missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABHD7 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5145425

 View more versions of this product

Product Code. 17900133

  • £389.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ABHD7 gene ontology (GO) annotation includes hydrolase activity and membrane.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ABHD7
Polyclonal
Unconjugated
EPHX4
ABHD7; abhydrolase domain containing 7; abhydrolase domain-containing protein 7; EH4; EPHX4; Ephxrp; epoxide hydrolase 4; Epoxide hydrolase-related protein; Gm1382
Rabbit
Antigen affinity chromatography, Protein A
RUO
253152, 384214
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Q6IE26, Q8IUS5
EPHX4
Recombinant Mouse EPHX4 protein, Lys36-Asp359 (Accession #Q6IE26).
200 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.