missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abhd5 Rabbit anti-Human, Rat, Clone: 2K8W9, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Abhd5 |
|---|---|
| Clone | 2K8W9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18397842
|
Novus Biologicals
NBP3-16675-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18376423
|
Bio-Techne
NBP3-16675-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Abhd5 Monoclonal antibody specifically detects Abhd5 in Human, Rat samples. It is validated for Western BlotSpecifications
| Abhd5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 51099 | |
| IgG | |
| Affinity purified |
| 2K8W9 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| abhydrolase domain containing 5, Abhydrolase domain-containing protein 5,1-acylglycerol-3-phosphate O-acyltransferase ABHD5, CDS, CGI58, EC 2.3.1.51, IECN2, Lipid droplet-binding protein CGI-58, MGC8731, NCIE2CGI-58 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Abhd5 (Q8WTS1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title