missing translation for 'onlineSavingsMsg'
Learn More

Abhd5 Rabbit anti-Human, Rat, Clone: 2K8W9, Novus Biologicals™

Product Code. 18376423 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376423 100 μg 100µL
18397842 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18376423 Supplier Bio-Techne Supplier No. NBP316675100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Abhd5 Monoclonal antibody specifically detects Abhd5 in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Abhd5
Applications Western Blot
Classification Monoclonal
Clone 2K8W9
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias abhydrolase domain containing 5, Abhydrolase domain-containing protein 5,1-acylglycerol-3-phosphate O-acyltransferase ABHD5, CDS, CGI58, EC 2.3.1.51, IECN2, Lipid droplet-binding protein CGI-58, MGC8731, NCIE2CGI-58
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Abhd5 (Q8WTS1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Lipid Droplets, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51099
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.