missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABHD2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | ABHD2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ABHD2 Polyclonal specifically detects ABHD2 in Human samples. It is validated for Western Blot.Specifications
| ABHD2 | |
| Polyclonal | |
| Rabbit | |
| P08910 | |
| 11057 | |
| Synthetic peptides corresponding to ABHD2(abhydrolase domain containing 2) The peptide sequence was selected from the middle region of ABHD2. Peptide sequence RESHTHRDRPSQQQPLRNQTTSSERRGEWEIQPSRQTNTSYLTSHLAADR. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| abhydrolase domain containing 2, abhydrolase domain-containing protein 2, alpha/beta hydrolase domain containing protein 2, EC 3.1.1, EC 3.1.1.-, HS1-2, LABH2PHPS1-2, Lung alpha/beta hydrolase 2, MGC111112, MGC26249, Protein PHPS1-2 | |
| ABHD2 | |
| IgG | |
| 48 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title