missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABHD2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-53199
This item is not returnable.
View return policy
Description
ABHD2 Polyclonal specifically detects ABHD2 in Human samples. It is validated for Western Blot.
Specifications
| ABHD2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| abhydrolase domain containing 2, abhydrolase domain-containing protein 2, alpha/beta hydrolase domain containing protein 2, EC 3.1.1, EC 3.1.1.-, HS1-2, LABH2PHPS1-2, Lung alpha/beta hydrolase 2, MGC111112, MGC26249, Protein PHPS1-2 | |
| Rabbit | |
| 48 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P08910 | |
| ABHD2 | |
| Synthetic peptides corresponding to ABHD2(abhydrolase domain containing 2) The peptide sequence was selected from the middle region of ABHD2. Peptide sequence RESHTHRDRPSQQQPLRNQTTSSERRGEWEIQPSRQTNTSYLTSHLAADR. | |
| Affinity purified | |
| RUO | |
| 11057 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction