missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £358.00
Specifications
| Antigen | ABCG4 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ABCG4 Polyclonal antibody specifically detects ABCG4 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ABCG4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| ABC Transporters, Lipid and Metabolism | |
| PBS with 50% glycerol, pH7.3. | |
| 64137 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ATP-binding cassette, sub-family G (WHITE), member 4, ATP-binding cassette, subfamily G, member 4, putative ABC transporter, WHITE2ATP-binding cassette sub-family G member 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 270-370 of human ABCG4 (NP_071452.2). KLYILSQGQCIFKGVVTNLIPYLKGLGLHCPTYHNPADFIIEVASGEYGDLNPMLFRAVQNGLCAMAEKKSSPEKNEVPAPCPPCPPEVDPIESHTFATST | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title