missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCC12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69063
This item is not returnable.
View return policy
Description
ABCC12 Polyclonal specifically detects ABCC12 in Mouse samples. It is validated for Western Blot.
Specifications
| ABCC12 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ATP-binding cassette transporter sub-family C member 12, ATP-binding cassette, sub-family C (CFTR/MRP), member 12, MGC27071, MRP9ATP-binding cassette sub-family C member 12, multidrug resistance-associated protein 9 | |
| Rabbit | |
| 153 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q80WJ6 | |
| Abcc12 | |
| Synthetic peptides corresponding to Abcc12 (ATP-binding cassette, sub-family C (CFTR/MRP), member 12) The peptide sequence was selected from the middle region of Abcc12. Peptide sequence NILFGEKYNHQRYQHTVHVCGLQKDLNSLPYGDLTEIGERGVNLSGGQRQ. | |
| Affinity purified | |
| RUO | |
| 94160 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction