missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ABCB11 Antibody (BSEP/7534), Novus Biologicals™
Mouse Monoclonal Antibody
£187.00 - £368.00
Specifications
| Antigen | ABCB11 |
|---|---|
| Clone | BSEP/7534 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043809
|
Novus Biologicals™
NBP3-26754-20ug |
20 μg |
£187.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043569
|
Novus Biologicals™
NBP3-26754-100ug |
100 μg |
£368.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ABCB11 Monoclonal antibody specifically detects ABCB11 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)Specifications
| ABCB11 | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Endocrinology, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 8647 | |
| IgG | |
| Protein A or G purified |
| BSEP/7534 | |
| Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ABC member 16, MDR/TAP subfamily, ABC16, ATP-binding cassette sub-family B member 11, ATP-binding cassette, sub-family B (MDR/TAP), member 11, bile salt export pump, BSEP, BSEPPFIC2, EC 3.6.3, EC 3.6.3.44, PFIC-2, PGY4, progressive familial intrahepatic cholestasis 2, sister p-glycoprotein, SPGPBRIC2 | |
| Recombinant fragment (around aa600-800) of human ABCB11 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title