missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ABCB11 Antibody (BSEP/7534), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-26754-20ug

 View more versions of this product

Product Code. 30043809

  • £187.00 / 20µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ABCB11 Monoclonal antibody specifically detects ABCB11 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ABCB11
Monoclonal
0.2 mg/mL
Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
ABC member 16, MDR/TAP subfamily, ABC16, ATP-binding cassette sub-family B member 11, ATP-binding cassette, sub-family B (MDR/TAP), member 11, bile salt export pump, BSEP, BSEPPFIC2, EC 3.6.3, EC 3.6.3.44, PFIC-2, PGY4, progressive familial intrahepatic cholestasis 2, sister p-glycoprotein, SPGPBRIC2
Recombinant fragment (around aa600-800) of human ABCB11 protein (exact sequence is proprietary)
20 μg
Cancer, Endocrinology, Signal Transduction
8647
Store at 4°C.
IgG
Western Blot, Immunohistochemistry (Paraffin)
BSEP/7534
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.