missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCA2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£186.00 - £375.00
Specifications
| Antigen | ABCA2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18673401
|
Novus Biologicals
NBP2-92339-0.02ml |
0.02 mL |
£186.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613411
|
Novus Biologicals
NBP2-92339-0.1ml |
0.1 mL |
£375.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ABCA2 Polyclonal antibody specifically detects ABCA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| ABCA2 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| ABC Transporters, Lipid and Metabolism | |
| PBS with 50% glycerol, pH7.3. | |
| 20 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ABC2ATP-binding cassette, sub-family A, member 2, ATP-binding cassette 2, ATP-binding cassette sub-family A member 2, ATP-binding cassette transporter 2, ATP-binding cassette, sub-family A (ABC1), member 2, EC 3.6.3, EC 3.6.3.41, KIAA1062, MGC129761 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 140-350 of human ABCA2 (NP_001597.2). SHLDRSTVSSFSLDSVARNPQELWRFLTQNLSLPNSTAQALLAARVDPPEVYHLLFGPSSALDSQSGLHKGQEPWSRLGGNPLFRMEELLLAPALLEQLTCTPGSGELGRILTVPESQKGALQGYRDAVCSGQAAARARRFSGLSAELRNQLDVAKVSQQLGLDAPNGSDSSPQAPPPRRLQALLGDLLDAQKVLQDVDVLSALALLLPQG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title