missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCA2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92339-0.1ml
This item is not returnable.
View return policy
Description
ABCA2 Polyclonal antibody specifically detects ABCA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| ABCA2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| ABC2ATP-binding cassette, sub-family A, member 2, ATP-binding cassette 2, ATP-binding cassette sub-family A member 2, ATP-binding cassette transporter 2, ATP-binding cassette, sub-family A (ABC1), member 2, EC 3.6.3, EC 3.6.3.41, KIAA1062, MGC129761 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 140-350 of human ABCA2 (NP_001597.2). SHLDRSTVSSFSLDSVARNPQELWRFLTQNLSLPNSTAQALLAARVDPPEVYHLLFGPSSALDSQSGLHKGQEPWSRLGGNPLFRMEELLLAPALLEQLTCTPGSGELGRILTVPESQKGALQGYRDAVCSGQAAARARRFSGLSAELRNQLDVAKVSQQLGLDAPNGSDSSPQAPPPRRLQALLGDLLDAQKVLQDVDVLSALALLLPQG | |
| 0.1 mL | |
| ABC Transporters, Lipid and Metabolism | |
| 20 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction