missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AATF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10282-1-AP-20UL
This item is not returnable.
View return policy
Description
AATF (apoptosis antagonizing transcription factor) was initially discovered as an interaction partner of ZIP kinase (ZIPK), a member of death-associated protein (DAP) kinase family of pro-apoptotic serine/threonine kinases. AATF is a phosphoprotein containing an acidic region and a putative leucine zipper domain and nuclear localization signal, features which are typical of transcription factors. AATF inhibits the ZIPK-mediated pro-apoptotic pathway and may activate other anti-apoptotic pathways. Recently, it has also been shown to protect neural cells against oxidative damage induced by amyloid b-peptide and to inhibit aberrant production of the beta-peptide by interacting with Par-4 (prostate apoptosis response-4), another pro-apoptotic leucine zipper protein that is associated with neuronal degeneration in Alzheimer′s disease (AD), suggesting that AATF may have potential therapeutic applications in both familial and sporadic forms of AD.Specifications
| AATF | |
| Polyclonal | |
| Unconjugated | |
| AATF | |
| AATF, CHE 1, CHE1, DED, Protein AATF, Rb binding protein Che 1, Traube | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114512, 26574, 56321 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9JKX4, Q9NY61, Q9QYW0 | |
| AATF | |
| AATF Fusion Protein Ag0195 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction