missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AARS2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92487-0.1ml
This item is not returnable.
View return policy
Description
AARS2 Polyclonal antibody specifically detects AARS2 in Human samples. It is validated for Western Blot
Specifications
| AARS2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| alanine tRNA ligase 2, mitochondrial (putative), alanine--tRNA ligase, alanyl-tRNA synthetase 2, mitochondrial (putative), alanyl-tRNA synthetase, mitochondrial, alaRS, bA444E17.1, COXPD8, KIAA1270, MTALARS, MT-ALARS, probable alanyl-tRNA synthetase, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 841-985 of human AARS2 (NP_065796.1). RRELLATVKMLQRRANTAIRKLQMGQAAKKTQELLERHSKGPLIVDTVSAESLSVLVKVVRQLCEQAPSTSVLLLSPQPMGKVLCACQVAQGAMPTFTAEAWALAVCSHMGGKAWGSRVVAQGTGSTTDLEAALSIAQTYALSQL | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 57505 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction