missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AARS2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | AARS2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18601242
|
Novus Biologicals
NBP2-92487-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18662292
|
Novus Biologicals
NBP2-92487-0.1ml |
0.1 mL |
£359.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AARS2 Polyclonal antibody specifically detects AARS2 in Human samples. It is validated for Western BlotSpecifications
| AARS2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| alanine tRNA ligase 2, mitochondrial (putative), alanine--tRNA ligase, alanyl-tRNA synthetase 2, mitochondrial (putative), alanyl-tRNA synthetase, mitochondrial, alaRS, bA444E17.1, COXPD8, KIAA1270, MTALARS, MT-ALARS, probable alanyl-tRNA synthetase, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 841-985 of human AARS2 (NP_065796.1). RRELLATVKMLQRRANTAIRKLQMGQAAKKTQELLERHSKGPLIVDTVSAESLSVLVKVVRQLCEQAPSTSVLLLSPQPMGKVLCACQVAQGAMPTFTAEAWALAVCSHMGGKAWGSRVVAQGTGSTTDLEAALSIAQTYALSQL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 57505 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title