missing translation for 'onlineSavingsMsg'
Learn More

AAK1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£134.00

Specifications

Antigen AAK1
Concentration 1 mg/mL
Dilution Western Blot 1-2 μg/mL, ELISA 1:100-1:2000
Applications Western Blot
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18586534
View Documents
Novus Biologicals
NBP1-76343-0.025mg
0.025 mg
£134.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AAK1 Polyclonal specifically detects AAK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

AAK1
Western Blot 1-2 μg/mL, ELISA 1:100-1:2000
Polyclonal
Rabbit
Protein Kinase
PBS with 0.02% Sodium Azide
Adaptor-associated kinase 1, AP2 associated kinase 1, AP2-associated protein kinase 1, DKFZp686F03202, DKFZp686K16132, EC 2.7.11.1, FLJ23712, FLJ25931, FLJ31060, FLJ45252, KIAA1048FLJ42882, MGC138170, MGC164568, MGC164570
AAK1
IgG
Affinity Purified
1 mg/mL
Western Blot
Unconjugated
RUO
Human
NP_055726
22848
Antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of the human Aak1. The immunogen is located within amino acids 830 - 880 of Aak1. Amino Acid Squence: PQRLPSQTESVTSNRTDS
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.