missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AAK1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76343-0.025mg
This item is not returnable.
View return policy
Description
AAK1 Polyclonal specifically detects AAK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| AAK1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Adaptor-associated kinase 1, AP2 associated kinase 1, AP2-associated protein kinase 1, DKFZp686F03202, DKFZp686K16132, EC 2.7.11.1, FLJ23712, FLJ25931, FLJ31060, FLJ45252, KIAA1048FLJ42882, MGC138170, MGC164568, MGC164570 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 | |
| NP_055726 | |
| AAK1 | |
| Antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of the human Aak1. The immunogen is located within amino acids 830 - 880 of Aak1. Amino Acid Squence: PQRLPSQTESVTSNRTDS | |
| 0.025 mg | |
| Protein Kinase | |
| 22848 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur