missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A2BP1 Mouse anti-Human, Mouse, Rat, Bovine, Clone: 1G10, Novus Biologicals™
Mouse Monoclonal Antibody
£176.00 - £372.00
Specifications
| Antigen | A2BP1 |
|---|---|
| Clone | 1G10 |
| Dilution | Western Blot 1:5000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18316702
|
Novus Biologicals
NBP3-11943-25UL |
25 μg |
£176.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18320615
|
Novus Biologicals
NBP3-11943-100UL |
100 μg |
£372.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A2BP1 Monoclonal antibody specifically detects A2BP1 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| A2BP1 | |
| Western Blot 1:5000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Bovine | |
| 2BP1, A2BP, A2BP1 Hexaribonucleotide-binding protein 1, Ataxin-2-binding protein 1, FOX-1, FOX1 hexaribonucleotide binding protein 1, Fox-1 homolog A, HRNBP1 ataxin 2-binding protein 1, RNA binding protein fox-1 homolog 1, RNA binding protein, fox-1 homolog (C. elegans) 1 | |
| Recombinant protein taken from the N-terminus of A2BP1 expressed in and purified from E. Coli. (UniProt: Q9NWB1) | |
| Primary | |
| Store at -20C. |
| 1G10 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Epigenetics, Neuroscience | |
| 100 ul in PBS | |
| 54715 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title