missing translation for 'onlineSavingsMsg'
Learn More

A2BP1 Mouse anti-Human, Mouse, Rat, Bovine, Clone: 1G10, Novus Biologicals™

Product Code. 18316702 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18316702 25 μg 25µL
18320615 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18316702 Supplier Novus Biologicals Supplier No. NBP31194325UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

A2BP1 Monoclonal antibody specifically detects A2BP1 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen A2BP1
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 1G10
Conjugate Unconjugated
Dilution Western Blot 1:5000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:500
Formulation 100 ul in PBS
Gene Alias 2BP1, A2BP, A2BP1 Hexaribonucleotide-binding protein 1, Ataxin-2-binding protein 1, FOX-1, FOX1 hexaribonucleotide binding protein 1, Fox-1 homolog A, HRNBP1 ataxin 2-binding protein 1, RNA binding protein fox-1 homolog 1, RNA binding protein, fox-1 homolog (C. elegans) 1
Host Species Mouse
Immunogen Recombinant protein taken from the N-terminus of A2BP1 expressed in and purified from E. Coli. (UniProt: Q9NWB1)
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 54715
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at -20C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.