missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ A.R.I.A Medium with 5% Horse Blood and Neomycin

Product Code. 15822437
Click to view available options
Description:
A.R.I.A Medium with 5% Horse Blood and Neomycin
Quantity:
10/Pk.
Manufacturing Origin:
United Kingdom
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 15822437

Brand: Thermo Scientific™ PB1244A

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Isolate fastidious and non‑fastidious anaerobic bacteria using a general purpose anaerobic growth medium.

Isolate fastidious and non‑fastidious anaerobic bacteria from clinical samples within 24 to 72 hours using Thermo Scientific™ Anaerobe Recovery and Isolation Agar (A.R.I.A™).

Made from Thermo Scientific™ Oxoid™ dehydrated culture media, A.R.I.A contains 5% defibrinated horse blood and peptones, carefully selected to support the growth of anaerobic bacteria. Neomycin is present as a selective agent for obligate anaerobes.

The following constituents have been added to the formulation to enhance performance:

  • Yeast extract provides a vitamin source, and starch is present to absorb any toxic metabolites
  • L-cysteine is present as a reducing agent and has shown to stimulate the growth of some anaerobes1
  • Glucose is the main carbohydrate, and sodium pyruvate is also added as an energy source for asaccharolytic cocci, such as Veillonella
  • Haemin and Vitamin K are also included as growth factors required by many Bacteriodes species2
  • Neomycin is present as a selective agent for obligate anaerobes

1. Shanson DC, Singh J. Effect of adding cysteine to brain heart infusion broth on the isolation of Bacteroides fragilis from experimental blood cultures. Journal of Clinical Pathology 1981; 34:221-3

2. Gibbons RJ, Macdonald JB. Haemin and vitamin K as required factors for cultivation of certain strains of B. melaninogevicus. Journal of Bacteriology 1960; 80:164-70
TRUSTED_SUSTAINABILITY

Specifications

Color Ruby red
Content And Storage 2° to 10°C, away from light
Description A.R.I.A Medium with 5% Horse Blood and Neomycin
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 18.5-20.5g
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.