missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A. aeolicus BPL/BioID2 Antibody (SS 3A5-E2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£174.00 - £402.00
Specifications
| Antigen | A. aeolicus BPL/BioID2 |
|---|---|
| Clone | SS 3A5-E2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1-2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18679516
|
Novus Biologicals
NBP2-59941-0.025mg |
0.025 mg |
£174.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620349
|
Novus Biologicals
NBP2-59941 |
0.1 mg |
£402.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A. aeolicus BPL/BioID2 Monoclonal antibody specifically detects A. aeolicus BPL/BioID2 in Bacteria samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| A. aeolicus BPL/BioID2 | |
| 1 mg/mL | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Epitope Tags | |
| PBS | |
| IgG1 κ | |
| Protein A purified |
| SS 3A5-E2 | |
| Western Blot 1:100 - 1-2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Bacteria | |
| GST fused to A. aeolicus BPL/BioID2 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title