missing translation for 'onlineSavingsMsg'
Learn More

A. aeolicus BPL/BioID2 Antibody (SS 3A5-E2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-59941-0.025mg

 View more versions of this product

Product Code. 18679516

  • £174.00 / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

A. aeolicus BPL/BioID2 Monoclonal antibody specifically detects A. aeolicus BPL/BioID2 in Bacteria samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

A. aeolicus BPL/BioID2
Monoclonal
1 mg/mL
Western Blot 1:100 - 1-2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500
Mouse
Protein A purified
RUO
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, Immunofluorescence
SS 3A5-E2
Unconjugated
PBS
GST fused to A. aeolicus BPL/BioID2
0.025 mg
Epitope Tags
Bacteria
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.