missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ A 83-01
GREENER_CHOICE

Product Code. p-4783704
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18084932

Brand: Tocris Bioscience™ 2939/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective inhibitor of TGF-βRI, ALK4 and ALK7

A 83-01 is a potent inhibitor of TGF-β type I receptor ALK5 kinase, type I activin/nodal receptor ALK4 and type I nodal receptor ALK7 (IC50 values are 12, 45 and 7.5 nM respectively). A 83-01 blocks phosphorylation of Smad2 and inhibits TGF-β-induced epithelial-to-mesenchymal transition. Only weakly inhibits ALK-1, -2, -3, -6 and MAPK activity. More potent than SB 431542 (Cat. No. 1614). A 83-01 inhibits differentiation of rat induced pluripotent stem cells (riPSCs) and increases clonal expansion efficiency. Helps maintain homogeneity and long-term in vitro self-renewal of human iPSCs. Also promotes neural differentiation of hPSCs as part of a chemical cocktail.

For more information about how A 83-01 may be used, see our protocol: Converting Fibroblasts into Cardiomyocytes (9C Cocktail)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 909910-43-6
Molecular Formula C25H19N5S

Specifications

Quantity 10 mg
Formula Weight 421.52
Purity >98%
Molecular Weight (g/mol) 421.522
Chemical Name or Material A 83-01
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.