missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 96-well V-Bottom PCR Microplates
GREENER_CHOICE

Product Code. 10156933 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Gray, White Bottom, White Bottom
White, Black Bottom, Black Bottom
White, Clear Bottom
Quantity:
240/Pk.
80/Pk.
Unit Size:
Pack of 240
Pack of 80
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity unitSize
10156933 White, Black Bottom, Black Bottom 240/Pk. Pack of 240
10136793 Gray, White Bottom, White Bottom 240/Pk. Pack of 240
10107223 White, Clear Bottom 80/Pk. Pack of 80
10146933 White, Black Bottom, Black Bottom 80/Pk. Pack of 80
10529203 White, Clear Bottom 240/Pk. Pack of 240
10248503 Gray, White Bottom, White Bottom 80/Pk. Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10156933 Supplier Eppendorf™ Supplier No. 0030605900

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Certified Yes
Format Standard
Detection Method Quantitative Protein Analysis
Array 8 x 12
Color White, Black Bottom, Black Bottom
Material Polycarbonate
No. of Wells 96
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) For PCR techniques
Max. Heat Stability 121°C
Quantity 240/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.