missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 96-Well U-Bottom PCR Microplates
GREENER_CHOICE

Product Code. 10444685 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Black
Gray, White Bottom
White, Black Bottom
White, Clear Bottom
Quantity:
240/Pk.
80/Pk.
Unit Size:
Pack of 240
Pack of 80
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity unitSize
10444685 Gray, White Bottom 240/Pk. Pack of 240
10763594 Black 240/Pk. Pack of 240
10218883 White, Clear Bottom 240/Pk. Pack of 240
10743424 White, Black Bottom 80/Pk. Pack of 80
10444495 Gray, White Bottom 80/Pk. Pack of 80
10668204 White, Clear Bottom 80/Pk. Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10444685 Supplier Eppendorf™ Supplier No. 0030605578

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Pipette, control and recover samples with this polypropylene microplate. Eppendorf™ 96-Well U-Bottom PCR Microplates are ideal for assays that require high resistance against temperature or solvents.

  • Unmatched, transparent polypropylene (PP) for improved sample visibility
  • OptiTrack™ Matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
  • RecoverMax™ well design: Optimized well geometry for minimal remaining volume and excellent mixing properties
  • Raised well rims and smooth surface for reliable closing, even with heat sealing
  • g-Safe™: exceptional centrifugation stability up to 6,000 × g
  • Polypropylene guarantees high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without slip agents, plasticizers or biocides – Materials which could have a negative effect on bioassays
  • Guaranteed functionality from -86 to +100°C
  • Autoclavable (121°C, 20 min.)
TRUSTED_SUSTAINABILITY

Specifications

Certified Yes
Format Standard
Detection Method Quantitative Protein Analysis
Array 8 x 12
Color Gray, White Bottom
Product Type 96-well PCR Microplate
Material Polypropylene
No. of Wells 96
Well Shape Round
Dnase Free Yes
For Use With (Application) For PCR techniques
Max. Heat Stability 121°C
Quantity 240/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.