missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ twin.tec™ PCR Plates 96-well, 150 μL - PCR Plates
GREENER_CHOICE

Product Code. 10492868 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Blue
Colorless
Skirt Style:
Skirted
Unskirted
Unskirted, Divisible
Product Type:
Divisible PCR Plates
Forensic DNA Grade PCR Plates
Low Profile PCR Plates
Microbiology PCR Plates
Packaging:
5 Bags x 4 Plates
Individually Blister Wrapped
Unit Size:
Pack of 10
Pack of 20
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Skirt Style Product Type Packaging unitSize
10492868 Colorless Skirted Microbiology PCR Plates Individually Blister Wrapped Pack of 10
10401728 Blue Skirted Microbiology PCR Plates Individually Blister Wrapped Pack of 10
10619779 Colorless Unskirted Low Profile PCR Plates 5 Bags x 4 Plates Pack of 20
10136287 Blue Unskirted Low Profile PCR Plates 5 Bags x 4 Plates Pack of 20
15655367 Colorless Skirted Forensic DNA Grade PCR Plates Individually Blister Wrapped Pack of 10
10309357 Colorless Unskirted, Divisible Divisible PCR Plates 5 Bags x 4 Plates Pack of 20
10679969 Blue Unskirted, Divisible Divisible PCR Plates 5 Bags x 4 Plates Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10492868 Supplier Eppendorf™ Supplier No. 0030129300
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. Raised well rims enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.

  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.

Certifications

Certified DNA-, DNase-, RNase-, and PCR-inhibitor-free.

† Certificate, test procedures and detailed information available upon request.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Human DNA, DNase, RNase-free
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging Individually Blister Wrapped
Color Colorless
Product Type Microbiology PCR Plates
Skirt Style Skirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 150 μL
No. of Wells 96
Sterility Non-sterile
For Use With (Application) RT-PCR applications
Max. RCF 2250 x g
Purity or Quality Grade PCR Clean and Free of Bacterial DNA (Non-sterile)
Quantity 10 Pk.
Rimmed Yes
Well Color Colorless
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.