Learn More
SPEX™ 96-Well Titer Plate with Square Wells
SPEX™ 96-Well Titer Plate features square wells that provide greater capacity than round-welled plates.
Brand: SPEX™ 2200
Code : Z3
Additional Details : Weight : 0.10000kg
Description
- Square-wells provide greater working capacity than typical round-well titer plates
- Extensively tested with various samples and shown to resist perforation by grinding media
- Alphanumeric marks for well identification
- Durable polypropylene construction
- Process 192 samples at a time by running two titer plates in the Standard Clamp Assembly
- Process 384 samples at a time by running four titer plates in the Large Capacity Clamp Assembly
- Deep-well titer plate with a 96-well format
- Each square well has a volume of 2.4 mL and a working volume of 2.0 mL
- To use, simply load each well with the sample, solution (if desired), and grinding media then seal with a 2201 Cap-Mat
- Place up to six titer plates in the Geno/Grinder™ for high-throughput tissue homogenization and cell lysis
Specifications
96-Well Plate | |
96 x 2.4 mL | |
Polypropylene |