Learn More
SPEX™ 96-Well Titer Plate with Round Wells
SPEX™ 96-Well Titer Plate features round wells constructed of durable polypropylene.
Brand: SPEX™ 2210-SPP
Code : Z3
Additional Details : Weight : 0.10000kg
Description
- Extensively tested with various samples and shown to resist perforation by grinding media
- Compatible with 2650 Cryo-Adapter to preserve temperature sensitive samples
- Alphanumeric marks for well identification
- Durable polypropylene construction
- Process 192 samples at a time by running two titer plates in the Standard Clamp Assembly
- Process 384 samples at a time by running four titer plates in the Large Capacity Clamp Assembly
- Deep-well titer plate with a 96-well format
- Each round-bottom well has a volume of 1.0 mL and a working volume of 0.7 mL
- To use, simply load each well with the sample, solution (if desired), and grinding media, then seal with a 2211 Cap-Mat. Place up to six titer plates in the Geno/Grinder™ for high-throughput tissue homogenization and cell lysis
Specifications
96-Well Plate | |
96 x 1 mL | |
Polypropylene |