missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 96-well tips, Clear

Product Code. 16600234 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
200 μL
250 μL
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Tip Style:
Low Retention, Maxymum Recovery
Standard
Unit Size:
Pack of 4800
This item is not returnable. View return policy

Product Code. 16600234

Brand: Axygen™ VT255LR

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Axygen™ Maxymum Recovery™ tips have been developed to work with the Agilent/Velocity11 and MGI Tech automation systems, which can be enabled for the high throughput of magnetic bead based RNA extraction of biological samples or pre-configured and qualified to automate high throughput pre-PCR samples.

  • Tip format: 96 tips
  • Tip volume range: Up to 250 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    • With or without aerosol-resistant filters
    • non-sterile or sterile
    • Axygen Maxymum Recovery surface
  • Compatibility:
    • Bravo automated liquid handling platform
    • VPrep™ vertical liquid handling pipetting station
    • MGISP-960
    • MGISP-100
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 250 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Format Rack, 96
Tip Style Low Retention, Maxymum Recovery
Quantity 96 Tips/Rk, 10 Racks/Pk., 5 Pk./Cs
For Use With (Equipment) Agilent Bravo and MGI Tech MGISP Automation Systems, VPrep Vertical Liquid Handling Pipetting Station
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.