missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 96-Well Semi-Skirted PCR Plates

Product Code. 12144122 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
Green
Natural
Red
Yellow
Unit Size:
Pack of 25
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
12144122 Yellow Pack of 25
11889261 Blue Pack of 25
12114122 Red Pack of 25
12104122 Natural Pack of 25
12134122 Green Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12144122

Please to purchase this item. Need a web account? Register with us today!

Directly compatible with all major thermal cyclers, including ABI instruments—can be used in ABI 96-well sequencers without adapters

  • Virgin polypropylene
  • Clear well bottom for sample visibility
  • Raised-rim design
  • Sealable with adhesive films and foils, heat seals, and cap strips
  • 300μL maximum volume when used with adhesive and heat seals
  • Cut-away corner and alphanumeric grid
  • Cleanroom produced

Specifications

Format Standard
Array 8 x 12
Color Yellow
Product Type PCR Microplate
Skirt Style Semi
Material Polypropylene
Volume (Metric) Well 300 μL
No. of Wells 96
For Use With (Application) PCR, RT-PCR applications
Rimmed Yes
Well Color Clear
Show More Show Less
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.