missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 96-Well Protein LoBind Deep Well Plates, Clear
GREENER_CHOICE

Product Code. 10228354 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 80
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10228354 Clear Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10228354 Supplier Eppendorf™ Supplier No. 0030509234_OLD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Combine a new well design with a sophisticated color coding system for ease of reading and orientation. The well geometry of Eppendorf™ 96-Well Protein LoBind Deep Well Plates allows high speed pipetting without splashing, excellent mixing and high speed centrifugation, which accelerates your manual and automated work processes.

  • Fulfill all current SBS, IVD, EN ISO 9001/2/3 and EN 13485 guidelines.
  • Recovermax - concaved round bottom well shape with smoothed edges
  • Optitrack - color framed plates with alphanumeric labelling
  • High chemical resistance
  • G-safe - highest centrifugation stability
  • LoBind - free of surface coating e.g., silicone
  • Free from DNA, DNase, RNase and PCR* inhibitor (PCR* clean)
  • LoBind Protein - minimum protein loss (less than 3% BSA 1μg/mL)
  • LoBind DNA - less than 1% loss of DNA/RNA at low concentrations and critical conditions (high salt, short DNA fragments)
  • Autoclavable
  • Optionally available with barcode
TRUSTED_SUSTAINABILITY

Specifications

Certified Yes
Format Standard
Detection Method Quantitative Protein Analysis
Array 8 x 12
Color Clear
Material Polycarbonate
Volume (Metric) Well 1000 μL
No. of Wells 96
Dnase Free Yes
For Use With (Application) For PCR techniques
Max. Heat Stability 121°C
Profile Deep well plates, 96 Wells
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.