Learn More
Fisherbrand™ 96 Well Plate, PP, U-Shape Well, Non Sterile, Chromatography tested
Shop All Fisherbrand ProductsSpecifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Material | Polypropylene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Total Volume (Metric) Well | 1200 μL |
| Well Shape | Round Bottom (U-well) |
| Height (Metric) | 40 mm |