missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 96-Well Flat Bottom PCR Microplates
GREENER_CHOICE

Product Code. 10298393 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Gray
White, Black
White, Clear Bottom
Quantity:
240/Pk.
80/Pk.
Unit Size:
Pack of 240
Pack of 80
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity unitSize
10298393 White, Clear Bottom 240/Pk. Pack of 240
15204526 White, Black 80/Pk. Pack of 80
10713224 White, Clear Bottom 80/Pk. Pack of 80
15294516 Gray 80/Pk. Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10298393 Supplier Eppendorf™ Supplier No. 0030605101

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For quantitative protein analysis

TRUSTED_SUSTAINABILITY

Specifications

Certified Yes
Format Standard
Detection Method Quantitative Protein Analysis
Array 8 x 12
Color White, Clear Bottom
Product Type 96-well PCR Microplate
Material Polypropylene
No. of Wells 96
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) For PCR techniques
Max. Heat Stability 121°C
Quantity 240/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.