missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uvex 9301 Ultravision Ventilated Goggles
Shop All Uvex Products
Click to view available options
:
Clear
Description
- Ergonomically designed full-vision goggles with a sophisticated ventilation system
- Suitable for workers who also require prescription eyeglasses
- Gray-transparent frame with clear, polycarbonate lens
- Lenses feature supravision plus coating
- Ideal protection for oil & gas industries
Supravision Plus Coating
- Inside: Anti-fog
- Outside: Anti-fog, scratch-resistant
- Suitable for environments with frequent temperature and humidity fluctuations
Specifications
Specifications
| Certifications/Compliance | EN 166:2001, EN 170:2002 |
| Lens Type | Supravision Plus |
| Lens Coating | Anti-fog |
| Color | Transparent |
| Lens Tint | Clear |
| Product Type | Anti-fog Goggles |
| For Use With (Application) | Oil and Gas Industries |
| Ventilation Details | Ventilated |
| Material | Polycarbonate |
| Disposable | Reusable |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction