Learn More
uvex 9193 Sportstyle Safety Spectacles
Product Code. p-7221900 Shop All Uvex Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Lens Coating:
Anti-fog
Anti-fog, Scratch-resistant
Lens Tint:
Clear
Gray
Light Brown
Light Green
Silver Mirror
Frame Color:
Black, Anthracite
Black, Blue
Black, Gray
Black, Lime Green
Black, White
White, Black
missing translation for 'unitSize':
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Coating Lens Tint Frame Color missing translation for 'unitSize'
17237291 Anti-fog Silver Mirror Black, Gray Each
15523427 Anti-fog, Scratch-resistant Clear Black, Blue Each
17217291 Anti-fog Clear Black, Blue Each
17207291 Anti-fog, Scratch-resistant Gray Black, White Each
17277281 Anti-fog, Scratch-resistant Light Brown White, Black Each
17297281 Anti-fog, Scratch-resistant Clear Black, Lime Green Each
17287281 Anti-fog, Scratch-resistant Clear Black, Gray Each
17227291 Anti-fog, Scratch-resistant Light Green Black, Anthracite Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 missing translation for 'options'
Product Code. 17237291 missing translation for 'mfr' uvex missing translation for 'supplierNo' 9193885
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Innovative safety spectacles featuring a sporty design, combined with maximum functionality and safety. Feather-light construction weighs only 23 grams, making these spectacles comfortable to wear for extended periods of time.
  • Innovative safety spectacles feature a sporty design that combines functionality and safety 
  • Feather-light construction weighs only 23 grams
  • Soft, adjustable nosepiece with soft, anti-slip earpieces for a pressure-free fit
  • Lens geometry ensures optimum vision
Supravision AR Coating
  • Inside: Anti-reflective
  • Outside: Anti-reflective
  • Ideal for working conditions with strong or mixed lighting conditions
Supravision Excellence Coating
  • Interior: Anti-fog
  • Exterior: Scratch-resistant and chemical-resistant
  • Lenses are easy to clean and less susceptible to soiling
Supravision Extreme Coating
  • Interior: Anti-fog
  • Exterior: Scratch-resistant
  • Ideal for working areas with long-term exposure to condensation, and high levels of dirt
Supravision Plus Coating
  • Inside: Anti-fog and scratch-resistant
  • Outside: Anti-fog and scratch-resistant
  • Suitable for environments with frequent temperature and humidity fluctuations
Supravision Variomatic Coating
  • Inside: Anti-fog
  • Outside: Scratch-resistant
  • Self-coloring filters automatically darken within 10 seconds and brighten after 30 seconds
  • Suitable for working areas with frequent movement from inside to outside, and from light to dark
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166:2001, EN 172:1994 + A1:2000 + A2:2001
Lens Coating Anti-fog
Lens Tint Silver Mirror
Product Type Ergonomic Safety Glasses
Frame Color Black, Gray
Model 9193 Sportstyle
Transmittance Value 0.12
Weight (Metric) 23 g
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard
Labeling W 166 FT CE - 5-3,1 W1 FTN CE