Learn More
uvex 9193 Sportstyle Safety Spectacles
Product Code. 17207291 Shop All Uvex Products
Click to view available options
Lens Coating:
Anti-fog
Anti-fog, Scratch-resistant
Lens Tint:
Clear
Gray
Light Brown
Light Green
Silver Mirror
Frame Color:
Black, Anthracite
Black, Blue
Black, Gray
Black, Lime Green
Black, White
White, Black
Unit Size:
Each
Product Code. 17207291
Brand: uvex 9193280
Only null left
Add to basket
Add to basket
Innovative safety spectacles featuring a sporty design, combined with maximum functionality and safety. Feather-light construction weighs only 23 grams, making these spectacles comfortable to wear for extended periods of time.
  • Innovative safety spectacles feature a sporty design that combines functionality and safety 
  • Feather-light construction weighs only 23 grams
  • Soft, adjustable nosepiece with soft, anti-slip earpieces for a pressure-free fit
  • Lens geometry ensures optimum vision
Supravision AR Coating
  • Inside: Anti-reflective
  • Outside: Anti-reflective
  • Ideal for working conditions with strong or mixed lighting conditions
Supravision Excellence Coating
  • Interior: Anti-fog
  • Exterior: Scratch-resistant and chemical-resistant
  • Lenses are easy to clean and less susceptible to soiling
Supravision Extreme Coating
  • Interior: Anti-fog
  • Exterior: Scratch-resistant
  • Ideal for working areas with long-term exposure to condensation, and high levels of dirt
Supravision Plus Coating
  • Inside: Anti-fog and scratch-resistant
  • Outside: Anti-fog and scratch-resistant
  • Suitable for environments with frequent temperature and humidity fluctuations
Supravision Variomatic Coating
  • Inside: Anti-fog
  • Outside: Scratch-resistant
  • Self-coloring filters automatically darken within 10 seconds and brighten after 30 seconds
  • Suitable for working areas with frequent movement from inside to outside, and from light to dark
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166:2001, EN 172:1994 + A1:2000 + A2:2001
Lens Type Supravision Extreme
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Gray
Product Type Ergonomic Safety Glasses
For Use With (Application) Construction, Demolition, and Machine Operation
Frame Color Black, White
Model 9193 Sportstyle
Transmittance Value 0.23
Weight (Metric) 23 g
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard
Labeling W 166 FT CE - 5-2.5 W 1 FTKN CE