Learn More
uvex 9192 Pheos Spectacles
Product Code. 17217281 Shop All Uvex Products
Click to view available options
Lens Coating:
Anti-fog
Anti-fog, Scratch-resistant
Lens Tint:
Clear
Gray
Unit Size:
Each
Product Code. 17217281
Brand: uvex 9192240
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Duo-spherical lenses provide a wide field of vision with soft, non-slip arm ends to prevent pressure points for enhanced comfort.
  • Soft, non-slip arm end prevents pressure points for enhanced comfort
  • Duo-spherical lenses provide a large field of vision
  • Design is extremely compatible with ear defenders, helmets, and bump caps
  • Eye Plus Protection provides additional eyebrow protection with a bevelled edge in the upper area
  • Easily add a headband and frame when working at heights or in confined spaces
Supravision Excellence Coating
  • Interior: Anti-fog
  • Exterior: Scratch-resistant and chemical-resistant
  • Lenses are easy to clean and less susceptible to soiling
Supravision Plus Coating
  • Inside: Anti-fog and scratch-resistant
  • Outside: Anti-fog and scratch-resistant
  • Suitable for environments with frequent temperature and humidity fluctuations
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166:2001, EN 170:2002
Lens Type Duo-Spherical
Lens Coating Anti-fog
Lens Tint Clear
Product Type Ergonomic Safety Glasses
Frame Color Black, Orange
Model 9192 Pheos
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard
Labeling W 166 F CE - 2C - 1,2 W 1 FKN CE