Learn More
Uvex™ 9161 Uncoated Safety Spectacles
Shop All Uvex Products
Click to view available options
:
Clear
Description
- Comfortable cover spectacle with frosted brow guard to block overhead glare
- Indirect ventilation due to special temple design with large side shields
- Uncoated
Specifications
Specifications
| Certifications/Compliance (Europe) | EN 166:2001, EN 170:2002 |
| Lens Tint | Clear |
| Frame Color | Clear |
| Lens Material | Polycarbonate |
| UV Protection | UV 400 |