Learn More
Uvex™ 9161 Coated Safety Spectacles
7.35 GBP valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Use promo code "21673" to get your promotional price.
Click to view available options
Clear
Description
- Comfortable cover spectacle with frosted brow guard to block overhead glare
- Indirect ventilation due to special temple design with large side shields
- Supravision plus coating is scratch-resistant and anti-fog on both sides
Specifications
Lens Type | Supravision Plus |
Certifications/Compliance (Europe) | EN 166:2001, EN 170:2002 |
Lens Tint | Clear |
Frame Color | Blue, Clear |
Lens Material | Polycarbonate |
UV Protection | UV 400 |