missing translation for 'onlineSavingsMsg'
Learn More

9-Phenanthrylmagnesium bromide, 0.25 M in 2-MeTHF, Thermo Scientific™

Product Code. 15451316
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15451316 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15451316 Supplier Thermo Scientific Alfa Aesar Supplier No. H54545.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
9-Phenanthrylmagnesium bromide is used as an organic chemical synthesis intermediate.

Solubility
It reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Specifications

CAS 71112-64-6
Molecular Formula C14H9BrMg
MDL Number MFCD00672008
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive; May form a precipitate
Synonym 9-phenanthrylmagnesium bromide, 9-phenanthryl-magnesiumbromid, magnesium,bromo-9-phenanthrenyl, magnesium,9h-phenanthren-9-ide,bromide, bromo phenanthren-9-yl magnesium, 9-phenanthrylmagnesium bromide, 0.5 m in thf, 9-phenanthrylmagnesium bromide, 0.25 m in 2-methf, 9-phenanthrylmagnesium bromide 0.5 m in tetrahydrofuran, grignard reagent
Solubility Information It reacts with water.
InChI Key ACMUZKQPXSHTGB-UHFFFAOYSA-M
SMILES Br[Mg]C1=CC2=C(C=CC=C2)C2=C1C=CC=C2
IUPAC Name bromo(phenanthren-9-yl)magnesium
Molecular Weight (g/mol) 281.44
PubChem CID 12396678
Formula Weight 281.43
Concentration or Composition (by Analyte or Components) 0.25M in 2-MeTHF
Chemical Name or Material 9-Phenanthrylmagnesium bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.