missing translation for 'onlineSavingsMsg'
Learn More

9-Hydroxy-3-(2-hydroxyethyl)-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, TRC

Product Code. 30389837
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389837 250 mg 250mg
30397518 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30389837 Supplier Toronto Research Chemicals Supplier No. TRCH943115250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181525-38-2
Molecular Formula C11 H12 N2 O3
Molecular Weight (g/mol) 220.22
Synonym 9-Hydroxy-3-(2-hydroxyethyl)-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, 3-(2-Hydroxyethyl)-2-methyl-9-hydroxy-4H-pyrido[1,2-a]pyrimidin-4-one, 3-(2-Hydroxyethyl)-9-hydroxy-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one
IUPAC Name 9-hydroxy-3-(2-hydroxyethyl)-2-methylpyrido[1,2-a]pyrimidin-4-one
SMILES CC1=C(CCO)C(=O)N2C=CC=C(O)C2=N1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C11H12N2O3/c1-7-8(4-6-14)11(16)13-5-2-3-9(15)10(13)12-7/h2-3,5,14-15H,4,6H2,1H3
Formula Weight 220.0848
Chemical Name or Material 9-Hydroxy-3-(2-hydroxyethyl)-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.