CAS RN 181525-38-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
9-Hydroxy-3-(2-hydroxyethyl)-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, TRC
CAS: 181525-38-2 Molecular Formula: C11 H12 N2 O3 Molecular Weight (g/mol): 220.22 Synonym: 9-Hydroxy-3-(2-hydroxyethyl)-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one,3-(2-Hydroxyethyl)-2-methyl-9-hydroxy-4H-pyrido[1,2-a]pyrimidin-4-one,3-(2-Hydroxyethyl)-9-hydroxy-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one IUPAC Name: 9-hydroxy-3-(2-hydroxyethyl)-2-methylpyrido[1,2-a]pyrimidin-4-one SMILES: CC1=C(CCO)C(=O)N2C=CC=C(O)C2=N1