missing translation for 'onlineSavingsMsg'
Learn More

(8S,9R,10S,11S,13S,14S)-9-Fluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,9,10,11,12,13,14,15-decahydro-3H-cyclopenta[a]phenanthren-3-one, TRC

Product Code. 30407979
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407979 1 mg
30408067 10 mg
30390135 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407979 Supplier Toronto Research Chemicals Supplier No. TRCF5918621MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61618-63-1
Molecular Formula C21H25FO4
Molecular Weight (g/mol) 360.42
Synonym Desoxymetasone Impurity A
SMILES C[C@]12C[C@H](O)[C@@]3(F)[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC=C2C(=O)CO

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C21H25FO4/c1-19-10-18(26)21(22)15(14(19)5-6-16(19)17(25)11-23)4-3-12-9-13(24)7-8-20(12,21)2/h6-9,14-15,18,23,26H,3-5,10-11H2,1-2H3/t14-,15-,18-,19-,20-,21-/m0/s1
Formula Weight 360.1737
Chemical Name or Material (8S,9R,10S,11S,13S,14S)-9-Fluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,9,10,11,12,13,14,15-decahydro-3H-cyclopenta[a]phenanthren-3-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.