CAS RN 61618-63-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(8S,9R,10S,11S,13S,14S)-9-Fluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,9,10,11,12,13,14,15-decahydro-3H-cyclopenta[a]phenanthren-3-one, TRC
CAS: 61618-63-1 Molecular Formula: C21H25FO4 Molecular Weight (g/mol): 360.42 Synonym: Desoxymetasone Impurity A SMILES: C[C@]12C[C@H](O)[C@@]3(F)[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC=C2C(=O)CO