missing translation for 'onlineSavingsMsg'
Learn More

(8E)-Dodecen-1-ol, TRC

Product Code. 30372858
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372858 100 mg 100mg
30388415 500 mg -
30383203 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30372858 Supplier Toronto Research Chemicals Supplier No. TRCD525705100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42513-42-8
Molecular Formula C12 H24 O
Molecular Weight (g/mol) 184.32
Synonym (8E)-8-Dodecen-1-ol, (E)-8-Dodecen-1-ol, (E)-8-Dodecenol, (E)-8-Dodecenol, (E)-8-Dodecenyl Alcohol
IUPAC Name (E)-dodec-8-en-1-ol
SMILES CCC\C=C\CCCCCCCO

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H24O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h4-5,13H,2-3,6-12H2,1H3/b5-4+
Formula Weight 184.1827
Chemical Name or Material (8E)-Dodecen-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.