CAS RN 42513-42-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(8E)-Dodecen-1-ol, TRC
CAS: 42513-42-8 Molecular Formula: C12 H24 O Molecular Weight (g/mol): 184.32 Synonym: (8E)-8-Dodecen-1-ol,(E)-8-Dodecen-1-ol,(E)-8-Dodecenol,(E)-8-Dodecenol,(E)-8-Dodecenyl Alcohol IUPAC Name: (E)-dodec-8-en-1-ol SMILES: CCC\C=C\CCCCCCCO