missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ 80nm Colloidal Gold Nanoparticles 15OD

Product Code. 18017653 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
Unit Size:
100mL
10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18017653 100 mL 100mL
18061554 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18017653 Supplier Novus Biologicals™ Supplier No. 38150100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ultra high quality with narrow size distribution and uniform shape

80 nm colloidal gold is an aqueous suspension of spherical metallic nanoparticles manufactured to the highest standard to ensure:
  • Uniform spherical shape
  • Narrow size distribution: 80 nm ± 8nm, <10% CV
  • Ultra high quality
  • Batch-to-batch consistency
  • High efficiency antibody binding for maximum sensitivity
  • Available at high concentrations to simplify assay development
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
Content And Storage 4°C Do not freeze.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.