Learn More
Agilent BioTek 800 TS Absorbance Reader
Shop All Agilent BioTek Products

Beschreibung
The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. Applications include ELISA, protein quantification, enzyme kinetics and cell-based assays
The color touch screen provides a visual user interface, making programming fast and intuitive. Expanded functionality and data analysis is available with Gen5 software. The 800 TS partners with the 50 TS washer, making a compact, versatile, and affordable system to automate many application workflows.
Warranty
12 months
Spezifikation
Spezifikation
| Product Type | Microplate Reader |
| Detector Type | Photodiode |
| Interface | USB Type B, USB Type A |
| Light Source | Tungsten Halogen Lamp |
| Reading Options | Endpoint, Kinetic, Well-area Scanning |
| Shaking | Linear (except NB configurations) |
| Software Type | Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option) |
| Bandwidth | 10 nm |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Filters | 340, 405, 450, 490, 630 nm |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
