missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy Amoxapine-d8, TRC

Product Code. 30388839
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388839 10 mg -
30407014 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30388839 Supplier Toronto Research Chemicals Supplier No. TRCH79780710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189505-24-5
Molecular Formula C17H8D8ClN3O2
Molecular Weight (g/mol) 337.83
Synonym 2-Chloro-11-(1-piperazinyl-d8)-dibenz[b,f][1,4]oxazepin-8-ol, 8-Hydroxyamoxapin-d8
SMILES [2H]C1(C(N(C(C(N1)([2H])[2H])([2H])[2H])C2=NC3=C(C=CC(=C3)O)OC4=C2C=C(C=C4)Cl)([2H])[2H])[2H]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H16ClN3O2/c18-11-1-3-15-13(9-11)17(21-7-5-19-6-8-21)20-14-10-12(22)2-4-16(14)23-15/h1-4,9-10,19,22H,5-8H2/i5D2,6D2,7D2,8D2
Formula Weight 337.1433
Chemical Name or Material 8-Hydroxy Amoxapine-d8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.