CAS RN 1189505-24-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
8-Hydroxy Amoxapine-d8, TRC
CAS: 1189505-24-5 Molecular Formula: C17H8D8ClN3O2 Molecular Weight (g/mol): 337.83 Synonym: 2-Chloro-11-(1-piperazinyl-d8)-dibenz[b,f][1,4]oxazepin-8-ol,8-Hydroxyamoxapin-d8 SMILES: [2H]C1(C(N(C(C(N1)([2H])[2H])([2H])[2H])C2=NC3=C(C=CC(=C3)O)OC4=C2C=C(C=C4)Cl)([2H])[2H])[2H]